Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF068707.1 | - | 8, 9 | 946 | ||
AK052384.1 | - | 8, 9 | 881 | |||
AK147177.1 | - | 9, 10 | 985 | |||
AK151844.1 | - | 9, 10 | 977 | |||
BC058402.1 | - | 7, 8 | 991 | |||
BC080798.1 | - | 8, 9 | 894 | |||
BC108374.1 | - | 9, 10 | 972 | 4936738 |