Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSN SIRNA Assays ›
Gene Symbol:
GSN
Gene Name:
gelsolin
Gene Aliases:
ADF, AGEL
Chromosome Location:
Chr.9: 121201082 - 121332844 on Build GRCh38
UniGene ID:
Hs.522373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6244
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000177.4
NP_000168.1
10
1484
NM_001127662.1
NP_001121134.1
10
1397
NM_001127663.1
NP_001121135.2
12
1602
NM_001127664.1
NP_001121136.1
12
1569
NM_001127665.1
NP_001121137.1
12
1564
NM_001127666.1
NP_001121138.1
12
1561
NM_001127667.1
NP_001121139.1
12
1552
NM_001258029.1
NP_001244958.1
10
1384
NM_001258030.1
NP_001244959.1
10
1461
NM_198252.2
NP_937895.1
11
1490
XM_005251943.1
XP_005252000.1
11
1588
XM_005251944.1
XP_005252001.1
11
1579
XM_005251945.3
XP_005252002.1
11
1492
XM_006717079.1
XP_006717142.1
11
1426
XM_011518584.1
XP_011516886.1
12
1579
XM_011518585.1
XP_011516887.1
10
1399
XM_011518586.1
XP_011516888.1
13
1602
XM_011518587.2
XP_011516889.1
13
1644
XM_011518588.2
XP_011516890.1
13
1606
XM_011518589.2
XP_011516891.1
13
1602
XM_011518590.2
XP_011516892.1
13
1554
XM_011518591.2
XP_011516893.1
18
2646
XM_011518592.1
XP_011516894.1
12
1468
XM_011518593.1
XP_011516895.1
13
1614
XM_011518594.1
XP_011516896.1
7
811
XM_017014643.1
XP_016870132.1
19
2797
XM_017014644.1
XP_016870133.1
19
2731
XM_017014645.1
XP_016870134.1
11
1543
XM_017014646.1
XP_016870135.1
14
1714
XM_017014647.1
XP_016870136.1
20
2930
XM_017014648.1
XP_016870137.1
13
1541
GenBank mRNA
AK092797.1
-
6
1625
AK096280.1
-
11
1463
AK125819.1
-
19
2293
AK295201.1
-
11
1328
AK295572.1
-
12
1523
AK297870.1
-
10
1342
AK299453.1
-
10
1449
AK300631.1
-
12
1536
AK304629.1
-
10
1405
AK304675.1
-
9
1438
AK307859.1
-
10
1348
AK315494.1
-
10
1455
BC026033.1
-
10
1428
4661084
BX647999.1
-
10
1386
X04412.1
-
10
1437
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146120, NM_001206367, NM_001206368, NM_001206369, AK150359, and others.
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis, engulfment
aging
positive regulation of gene expression
oligodendrocyte development
striated muscle atrophy
extracellular matrix disassembly
actin filament polymerization
protein destabilization
tissue regeneration
sequestering of actin monomers
cellular protein metabolic process
actin nucleation
response to ethanol
negative regulation of viral entry into host cell
phosphatidylinositol-mediated signaling
actin filament severing
barbed-end actin filament capping
positive regulation of actin nucleation
response to folic acid
actin filament capping
cilium morphogenesis
cellular response to cadmium ion
regulation of podosome assembly
actin filament reorganization
renal protein absorption
hepatocyte apoptotic process
positive regulation of keratinocyte apoptotic process
regulation of wound healing, spreading of epidermal cells
regulation of establishment of T cell polarity
regulation of plasma membrane raft polarization
regulation of receptor clustering
positive regulation of protein processing in phagocytic vesicle
amyloid fibril formation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
actin binding
calcium ion binding
protein binding
protein domain specific binding
myosin II binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0