Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290788.1 | NP_001277717.1 | 16 | 1607 | ||
NM_009702.3 | NP_033832.2 | 16 | 1607 | |||
GenBank mRNA | AK082873.1 | - | 16 | 1547 | ||
AK122310.1 | - | 16 | 1595 | |||
AK138700.1 | - | 16 | 1663 | |||
AK145547.1 | - | 16 | 1592 | |||
AK151716.1 | - | 16 | 1578 | |||
BC042479.1 | - | 16 | 1587 | 5363276 | ||
U90333.1 | - | 16 | 1455 |