Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009703.2 | NP_033833.1 | 12 | 1407 | ||
GenBank mRNA | AK034861.1 | - | 13 | 1514 | ||
AK085823.1 | - | 12 | 1373 | |||
BC004757.1 | - | 12 | 1363 | 3154293 | ||
D00024.1 | - | 7 | 769 |