Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301424.1 | NP_001288353.1 | 10 | 1492 | ||
NM_007513.4 | NP_031539.3 | 10 | 1608 | |||
GenBank mRNA | AK139342.1 | - | 10 | 1584 | ||
AK147991.1 | - | 10 | 1517 | |||
AK163202.1 | - | 10 | 1492 | |||
BC068234.1 | - | 10 | 1499 | |||
BC145779.1 | - | 9 | 1413 | 40131082 | ||
BC145781.1 | - | 9 | 1413 | 40131084 | ||
JN226407.1 | - | 8 | 1328 | |||
JN226408.1 | - | 8 | 1328 | |||
JN226410.1 | - | 8 | 1328 | |||
M26687.1 | - | 10 | 1526 |