Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044740.2 | NP_001038205.1 | 2 | 504 | ||
NM_007514.3 | NP_031540.2 | 2 | 504 | |||
GenBank mRNA | AK154621.1 | - | 2 | 478 | ||
AK155558.1 | - | 2 | 479 | |||
AK171369.1 | - | 2 | 476 | |||
BB648063.1 | - | 2 | 478 | |||
BC021653.1 | - | 2 | 498 | |||
BC127082.1 | - | 2 | 420 | 40111372 | ||
CN710433.1 | - | 1 | 111 | |||
DQ086834.1 | - | 2 | 459 | |||
L03290.1 | - | 1 | 306 | |||
L11600.1 | - | 1 | 306 | |||
L29006.1 | - | 1 | 829 | |||
M62838.1 | - | 2 | 417 |