Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301649.1 | NP_001288578.1 | 6 | 800 | ||
NM_007517.4 | NP_031543.2 | 7 | 951 | |||
GenBank mRNA | AK154707.1 | - | 7 | 820 | ||
AK158035.1 | - | 7 | 742 | |||
AK167335.1 | - | 7 | 741 | |||
AK171011.1 | - | 7 | 754 | |||
AK171184.1 | - | 7 | 747 | |||
AK171202.1 | - | 7 | 761 | |||
AK171249.1 | - | 7 | 741 | |||
AK171541.1 | - | 7 | 741 | |||
AK171621.1 | - | 7 | 734 | |||
BC012699.1 | - | 6 | 584 | 3986517 | ||
BC016485.1 | - | 7 | 744 | 3962982 | ||
U41736.1 | - | 7 | 749 |