Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009740.2 | NP_033870.1 | 3 | 598 | ||
GenBank mRNA | AF057701.1 | - | 3 | 482 | ||
AF100339.1 | - | 3 | 490 | |||
AF127387.1 | - | 3 | 401 | |||
AF134396.1 | - | 3 | 482 | |||
AJ006289.1 | - | 3 | 479 | |||
AK076082.1 | - | 3 | 510 | |||
AK140179.1 | - | 3 | 470 | |||
AK146236.1 | - | 3 | 504 | |||
AK150883.1 | - | 3 | 499 | |||
AK152563.1 | - | 3 | 499 | |||
AK152847.1 | - | 3 | 495 | |||
AK156890.1 | - | 3 | 552 | |||
AK168084.1 | - | 3 | 504 | |||
AK169126.1 | - | 3 | 505 | |||
AK172158.1 | - | 3 | 509 | |||
AK207365.1 | - | 2 | 90 | |||
BC003703.1 | - | 3 | 517 | |||
BC024379.1 | - | 3 | 476 | 4976147 | ||
BC172007.1 | - | 3 | 482 | 9054293 | ||
BC172009.1 | - | 3 | 482 | 9054295 | ||
BU707261.1 | - | 3 | 598 |