Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007535.2 | NP_031561.2 | 1 | 289 | ||
GenBank mRNA | BC120720.1 | - | 1 | 289 | 40129643 | |
BC132545.1 | - | 1 | 289 |
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 1 | 399 | ||
GenBank mRNA | BC028762.1 | - | 345 | 1226745 | ||
CT010250.1 | - | 268 | ||||
L16462.1 | - | 1 | 399 |
Gene Symbol: Bcl2a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007534.3 | NP_031560.2 | 1 | 355 | ||
GenBank mRNA | AK138369.1 | - | 1 | 335 | ||
AK152200.1 | - | 1 | 325 | |||
BC100462.1 | - | 1 | 332 | 30295442 | ||
BC116241.1 | - | 1 | 355 | 40091848 | ||
BE650553.1 | - | 1 | 449 |
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007536.2 | NP_031562.1 | 1 | 399 | ||
GenBank mRNA | AK165576.1 | - | 1 | 151 | ||
BC027536.1 | - | 1 | 312 | 1363928 | ||
BC053965.1 | - | 1 | 421 | |||
BC116240.2 | - | 1 | 355 | 40091847 |