Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009746.2 | NP_033876.1 | 3 | 507 | ||
GenBank mRNA | AK131935.1 | - | 3 | 396 | ||
BC005673.1 | - | 3 | 507 | 3153237 | ||
BC058530.1 | - | 3 | 378 | 6818556 | ||
Y11905.1 | - | 4 | 325 |