Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042527.2 | NP_001035992.1 | 16 | 3166 | ||
NM_007550.4 | NP_031576.4 | 16 | 3437 | |||
GenBank mRNA | AB008674.1 | - | 16 | 3427 | ||
AK137022.1 | - | 16 | 3149 | |||
AK139975.1 | - | 1 | 2043 | |||
Z98263.1 | - | 16 | 3144 |