Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256019.1 | NP_001242948.1 | 4 | 1392 | ||
NM_007558.3 | NP_031584.1 | 4 | 1392 | |||
GenBank mRNA | AK157978.1 | - | 4 | 1054 | ||
BC052168.1 | - | 4 | 1390 | 6334507 | ||
M97017.1 | - | 4 | 940 |