Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009767.2 | NP_033897.1 | 5, 6 | 744 | ||
GenBank mRNA | AK034427.1 | - | 5, 6 | 717 | ||
AK135974.1 | - | 5, 6 | 696 | |||
BC132322.1 | - | 5, 6 | 663 | 40130599 | ||
BC138730.1 | - | 5, 6 | 663 | 8861752 |