Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077184.1 | NP_001070652.1 | 4 | 556 | ||
NM_009768.2 | NP_033898.1 | 5 | 904 | |||
GenBank mRNA | AK002332.1 | - | 4 | 408 | ||
AK011364.1 | - | 4 | 407 | |||
AK147225.1 | - | 4 | 5230 | |||
AK207023.1 | - | 2 | 155 | |||
AK208329.1 | - | 2 | 156 | |||
AK209680.1 | - | 2 | 156 | |||
AK215038.1 | - | 2 | 156 | |||
AK216955.1 | - | 2 | 156 | |||
AY089967.1 | - | 5 | 762 | |||
BC010270.1 | - | 4 | 399 | 3589236 | ||
D00611.1 | - | 4 | 406 | |||
S63813.1 | - | 4 | 406 | |||
Y16256.1 | - | 4 | 406 | |||
Y16257.1 | - | 3 | 402 | |||
Y16258.1 | - | 3 | 348 |