Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147778.3 | NP_680087.1 | 6 | 454 | ||
GenBank mRNA | AK007318.1 | - | 6 | 459 | ||
AK041634.1 | - | 6 | 455 | |||
AK145955.1 | - | 6 | 482 | |||
AK145999.1 | - | 6 | 484 | |||
BC038077.1 | - | 6 | 496 | 4013160 | ||
BC048815.1 | - | 6 | 465 | 3482368 |