Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007573.2 | NP_031599.2 | 4 | 617 | ||
GenBank mRNA | AF300619.1 | - | 4 | 556 | ||
AJ001101.1 | - | 4 | 562 | |||
AK010601.1 | - | 4 | 555 | |||
AK010746.1 | - | 4 | 581 | |||
AK011868.1 | - | 4 | 608 | |||
AK169764.1 | - | 4 | 600 | |||
BC038075.1 | - | 4 | 601 | 3154276 |