Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009779.2 | NP_033909.1 | 2 | 657 | ||
GenBank mRNA | AK054017.1 | - | 2 | 660 | ||
AK154134.1 | - | 2 | 583 | |||
AK154229.1 | - | 2 | 645 | |||
AK154609.1 | - | 2 | 584 | |||
AY786554.1 | - | 1 | 475 | |||
BC003728.1 | - | 2 | 570 | 3593758 | ||
U77460.1 | - | 2 | 505 |
Set Membership: |
G-protein coupled receptor |