Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042725.1 | NP_001036190.1 | 8, 9 | 875 | ||
NM_007588.2 | NP_031614.2 | 8, 9 | 1124 | |||
GenBank mRNA | AK039161.1 | - | 8, 9 | 1069 | ||
AK133983.1 | - | 8, 9 | 876 | |||
AK138275.1 | - | 8, 9 | 1126 | |||
AK162391.1 | - | 8, 9 | 879 | |||
BC119232.1 | - | 6, 7 | 631 | 8733981 | ||
BC119272.1 | - | 6, 7 | 631 | 8734021 | ||
U18542.1 | - | 8, 9 | 1026 |
Set Membership: |
![]() |