Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 12, 13 | 1593 | ||
NM_007598.4 | NP_031624.2 | 12, 13 | 1577 | |||
GenBank mRNA | AK053351.1 | - | 12, 13 | 1401 | ||
AK128889.1 | - | 12, 13 | 1399 | |||
AK137222.1 | - | 12, 13 | 1382 | |||
AK144094.1 | - | 12, 13 | 1385 | |||
AK154002.1 | - | 12, 13 | 1597 | |||
AK156556.1 | - | 12, 13 | 1411 | |||
AK158150.1 | - | 11, 12 | 2241 | |||
AK165565.1 | - | 12, 13 | 1380 | |||
AK169652.1 | - | 12, 13 | 1372 | |||
AK170901.1 | - | 12, 13 | 1388 | |||
AK171388.1 | - | 12, 13 | 1395 | |||
BC005446.1 | - | 11, 12 | 1376 | 3600514 | ||
BC005472.1 | - | 11, 12 | 1369 | 3590890 | ||
L12367.1 | - | 12, 13 | 1403 |