Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037761.2 | NP_001032850.1 | 8 | 816 | ||
NM_001271405.1 | NP_001258334.1 | 8 | 795 | |||
NM_001271406.1 | NP_001258335.1 | 8 | 781 | |||
NM_009798.4 | NP_033928.1 | 8 | 816 | |||
GenBank mRNA | AK153367.1 | - | 8 | 593 | ||
AK153394.1 | - | 8 | 593 | |||
AK156778.1 | - | 8 | 785 | |||
AK159390.1 | - | 8 | 790 | |||
AK160082.1 | - | 8 | 791 | |||
AK162757.1 | - | 8 | 783 | |||
AK168738.1 | - | 8 | 744 | |||
AK177260.1 | - | 3 | 144 | |||
AK177711.1 | - | 3 | 143 | |||
AK179503.1 | - | 3 | 144 | |||
AK182558.1 | - | 3 | 143 | |||
AK182951.1 | - | 3 | 145 | |||
AK186717.1 | - | 3 | 144 | |||
AK187793.1 | - | 3 | 144 | |||
AK188337.1 | - | 3 | 144 | |||
AK192076.1 | - | 3 | 144 | |||
AK198394.1 | - | 4 | 234 | |||
AK200176.1 | - | 3 | 144 | |||
AK207588.1 | - | 3 | 144 | |||
AK212957.1 | - | 3 | 207 | |||
AK219201.1 | - | 3 | 144 | |||
BC002053.1 | - | 8 | 758 | 3590980 | ||
FJ692320.1 | - | 8 | 781 | |||
U10406.1 | - | 8 | 736 | |||
U10407.1 | - | 8 | 707 |