Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042072.1 | - | 11 | 1315 | ||
AK133829.1 | - | 11 | 1326 | |||
AK142569.1 | - | 4 | 454 | |||
AK170649.1 | - | 10 | 1048 | |||
AK184247.1 | - | 2 | 93 | |||
AK191150.1 | - | 2 | 93 | |||
AK193297.1 | - | 2 | 93 | |||
AK194917.1 | - | 125 | ||||
AK205872.1 | - | 2 | 93 | |||
AK208643.1 | - | 2 | 93 | |||
AK210856.1 | - | 2 | 93 | |||
BC034262.1 | - | 11 | 1381 | 4460089 | ||
D28492.1 | - | 11 | 1304 | |||
Y13085.1 | - | 11 | 1383 |