Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111026.2 | NP_001104496.1 | 5 | 1044 | ||
NM_001111027.2 | NP_001104497.1 | 6 | 1238 | |||
NM_009822.3 | NP_033952.1 | 5 | 976 | |||
GenBank mRNA | AB221624.1 | - | 4 | 623 | ||
AK032132.1 | - | 5 | 911 | |||
AK142006.1 | - | 5 | 1045 | |||
BC139200.1 | - | 5 | 702 | 8862222 | ||
BC139201.1 | - | 5 | 706 | 8862223 | ||
D32007.1 | - | 5 | 976 | |||
X79989.1 | - | 5 | 683 |