Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271353.1 | NP_001258282.1 | 10 | 1204 | ||
NM_144855.3 | NP_659104.1 | 10 | 1207 | |||
NM_178224.3 | NP_835742.1 | 10 | 1207 | |||
GenBank mRNA | AK077669.1 | - | 10 | 1168 | ||
AK085692.1 | - | 10 | 1171 | |||
BC013472.1 | - | 10 | 1133 | 4221477 | ||
BC013480.1 | - | 10 | 1143 | 4239757 | ||
BC022144.1 | - | 10 | 1168 | |||
BC026595.1 | - | 10 | 1140 | 4974777 |