Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007634.4 | NP_031660.3 | 7 | 750 | ||
GenBank mRNA | AK030298.1 | - | 689 | |||
AK031613.1 | - | 7 | 690 | |||
AK032961.1 | - | 7 | 676 | |||
AK082485.1 | - | 7 | 718 | |||
AK085183.1 | - | 7 | 687 | |||
AK159594.1 | - | 7 | 751 | |||
AK162447.1 | - | 7 | 690 | |||
AK169263.1 | - | 7 | 701 | |||
BC021658.1 | - | 7 | 670 | |||
BC037662.1 | - | 7 | 702 | 5353169 | ||
U20612.1 | - | 7 | 688 | |||
U20636.1 | - | 7 | 805 | |||
Z47766.1 | - | 7 | 690 |