Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GUK1 SIRNA Assays ›
Gene Symbol:
GUK1
Gene Name:
guanylate kinase 1
Gene Aliases:
GMK
Chromosome Location:
Chr.1: 228139643 - 228148955 on Build GRCh38
UniGene ID:
Hs.376933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6357
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GUK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000858.5
NP_000849.1
3, 4
408
NM_001159390.1
NP_001152862.1
2, 3
243
NM_001159391.1
NP_001152863.1
2, 3
157
NM_001242839.1
NP_001229768.1
2, 3
183
NM_001242840.1
NP_001229769.1
2, 3
243
XM_005273103.3
XP_005273160.1
4, 5
579
XM_005273104.4
XP_005273161.1
4, 5
577
XM_005273105.3
XP_005273162.1
3, 4
436
XM_005273106.2
XP_005273163.1
2, 3
624
XM_011544174.2
XP_011542476.1
2, 3
3007
XM_017001091.1
XP_016856580.1
4, 5
580
XM_017001092.1
XP_016856581.1
3, 4
915
XM_017001093.1
XP_016856582.1
3, 4
375
XM_017001094.1
XP_016856583.1
2, 3
213
XM_017001095.1
XP_016856584.1
3, 4
372
XM_017001096.1
XP_016856585.1
2, 3
212
GenBank mRNA
AK303845.1
-
2, 3
196
BC006249.2
-
3, 4
346
3946293
BC007369.2
-
1, 2
60
BC009914.2
-
3, 4
320
2821563
BC091480.1
-
2, 3
163
30340102
BE780558.1
-
2, 3
130
BI771824.1
-
2, 3
183
BM790067.1
-
2, 3
283
BU162698.1
-
224
BU194785.1
-
209
BU931096.1
-
200
CR456836.1
-
1, 2
105
CR542006.1
-
1, 2
105
L76200.1
-
1, 2
105
U66895.1
-
1, 2
111
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00176133_m1
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide metabolic process
dGDP biosynthetic process
nucleobase-containing small molecule interconversion
GDP-mannose metabolic process
glycoprotein transport
ATP metabolic process
GMP metabolic process
dGMP metabolic process
dATP metabolic process
GDP biosynthetic process
nucleotide phosphorylation
guanylate kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0