Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007649.4 | NP_031675.1 | 2 | 189 | ||
GenBank mRNA | AB196820.1 | - | 2 | 150 | ||
AB196821.1 | - | 2 | 150 | |||
AB196822.1 | - | 2 | 150 | |||
AB196823.1 | - | 2 | 150 | |||
AK007741.1 | - | 2 | 190 | |||
AK084984.1 | - | 2 | 218 | |||
AK088027.1 | - | 2 | 236 | |||
AK151601.1 | - | 2 | 180 | |||
BC060977.1 | - | 2 | 217 | 30296173 | ||
X17501.1 | - | 2 | 200 | |||
X53526.1 | - | 2 | 200 |