Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033122.3 | NP_001028294.1 | 3 | 349 | ||
GenBank mRNA | AK017979.1 | - | 3 | 350 | ||
AK151076.1 | - | 3 | 350 | |||
AK153224.1 | - | 3 | 350 | |||
BC106997.2 | - | 3 | 334 | |||
BC128272.1 | - | 3 | 334 | 40081252 | ||
BC128273.1 | - | 3 | 334 | 40081255 | ||
BC139135.1 | - | 3 | 311 | 8862157 | ||
BC139136.1 | - | 3 | 311 | 8862158 |