Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141737.1 | - | 9 | 961 | ||
AK181318.1 | - | 6 | 355 | |||
BC004669.1 | - | 8 | 665 | 3498846 | ||
L16926.1 | - | 8 | 1143 | |||
U15562.1 | - | 9 | 927 |