Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243769.1 | NP_001230698.1 | 4 | 369 | ||
NM_009861.3 | NP_033991.1 | 4 | 369 | |||
GenBank mRNA | AK003098.1 | - | 4 | 290 | ||
AK051543.1 | - | 4 | 290 | |||
AK075567.1 | - | 4 | 288 | |||
AK144216.1 | - | 4 | 292 | |||
AK145659.1 | - | 3 | 133 | |||
AK151087.1 | - | 4 | 314 | |||
AK151726.1 | - | 4 | 291 | |||
AK153564.1 | - | 4 | 313 | |||
AK154870.1 | - | 4 | 288 | |||
AK159470.1 | - | 4 | 331 | |||
AK166281.1 | - | 4 | 288 | |||
AK167195.1 | - | 4 | 277 | |||
AK167400.1 | - | 4 | 282 | |||
AK167609.1 | - | 4 | 287 | |||
AK168013.1 | - | 3 | 282 | |||
AK168076.1 | - | 4 | 282 | |||
AK168089.1 | - | 3 | 282 | |||
AK168276.1 | - | 4 | 282 | |||
AK168503.1 | - | 4 | 303 | |||
AK168758.1 | - | 4 | 322 | |||
AK168820.1 | - | 4 | 295 | |||
AK169122.1 | - | 4 | 303 | |||
AK169232.1 | - | 4 | 282 | |||
AK169805.1 | - | 4 | 307 | |||
CD551112.1 | - | 4 | 356 | |||
L11318.1 | - | 3 | 205 | |||
L78075.1 | - | 3 | 209 | |||
U37720.1 | - | 4 | 277 |