Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161623.1 | NP_001155095.1 | 10 | 774 | ||
NM_009862.2 | NP_033992.2 | 12 | 1151 | |||
GenBank mRNA | AF081536.1 | - | 12 | 1151 | ||
AF081537.1 | - | 11 | 850 | |||
AF081538.1 | - | 11 | 850 | |||
AF098068.1 | - | 11 | 897 | |||
AJ223729.1 | - | 11 | 871 | |||
AK147013.1 | - | 11 | 868 | |||
AK167850.1 | - | 10 | 762 | |||
BC006856.1 | - | 11 | 866 | |||
BC028635.1 | - | 11 | 904 | 1262927 |