Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021957.3 | NP_068776.2 | 16 | 2282 | ||
XM_006719063.3 | XP_006719126.1 | 15 | 1814 | |||
GenBank mRNA | BC126310.1 | - | 16 | 2052 | 8992026 | |
BC126312.1 | - | 16 | 2052 | 8992028 | ||
D29685.1 | - | 16 | 2284 | |||
S70004.1 | - | 16 | 2055 |