Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111099.1 | NP_001104569.1 | 3 | 763 | ||
NM_007669.4 | NP_031695.1 | 3 | 737 | |||
GenBank mRNA | AK007630.1 | - | 3 | 735 | ||
AK089093.1 | - | 3 | 735 | |||
AK139306.1 | - | 3 | 738 | |||
AK149910.1 | - | 3 | 738 | |||
AK170467.1 | - | 3 | 738 | |||
BC002043.1 | - | 3 | 690 | 3495942 | ||
U09507.1 | - | 2 | 835 |