Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007687.5 | NP_031713.1 | 3, 4 | 542 | ||
GenBank mRNA | AK013943.1 | - | 3, 4 | 471 | ||
AK020471.1 | - | 3, 4 | 513 | |||
AK050651.1 | - | 473 | ||||
AK088633.1 | - | 3, 4 | 478 | |||
AK088793.1 | - | 3, 4 | 477 | |||
AK167795.1 | - | 3, 4 | 473 | |||
BC046225.1 | - | 3, 4 | 437 | 5691650 | ||
BC058726.1 | - | 3, 4 | 470 | 5715707 | ||
BC094357.1 | - | 3, 4 | 452 | 6820355 | ||
CX218255.1 | - | 3, 4 | 578 | |||
D00472.1 | - | 3, 4 | 491 |