Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007692.6 | NP_031718.1 | 10 | 1420 | ||
GenBank mRNA | AB011000.1 | - | 10 | 1392 | ||
AB011001.1 | - | 6 | 1947 | |||
AK078101.1 | - | 10 | 1414 | |||
AK170077.1 | - | 9 | 1101 | |||
AK177995.1 | - | 4 | 354 | |||
AK187259.1 | - | 3 | 251 | |||
BC068195.1 | - | 10 | 938 | 4164116 | ||
BC069925.1 | - | 8 | 1741 |