Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007694.4 | NP_031720.1 | 4 | 1824 | ||
GenBank mRNA | AK077487.1 | - | 4 | 1393 | ||
AK077489.1 | - | 4 | 1432 | |||
AK133629.1 | - | 4 | 1429 | |||
AK133739.1 | - | 4 | 1433 | |||
AK133786.1 | - | 4 | 1434 | |||
AK158857.1 | - | 4 | 1825 | |||
BC014736.1 | - | 4 | 1425 | 4511248 | ||
X51429.1 | - | 4 | 1420 | |||
X53028.1 | - | 4 | 1430 |