Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163432.1 | NP_001156904.1 | 7 | 1050 | ||
NM_007712.3 | NP_031738.2 | 7 | 1053 | |||
GenBank mRNA | AF033564.1 | - | 7 | 775 | ||
AK134849.1 | - | 1 | 1294 | |||
AK148377.1 | - | 5 | 2081 | |||
AK161259.1 | - | 3 | 1225 | |||
BC015080.1 | - | 7 | 1038 | 3995512 | ||
CN538691.1 | - | 2 | 215 |