Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016706.2 | NP_057915.2 | 1, 2 | 276 | ||
GenBank mRNA | AK146366.1 | - | 1, 2 | 263 | ||
BC031167.1 | - | 1, 2 | 294 | 4983919 | ||
BY065340.1 | - | 1, 2 | 277 |