Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 50 | 3699 | ||
GenBank mRNA | AK075707.1 | - | 28 | 2272 | ||
AK132496.1 | - | 50 | 3695 | |||
AK159120.1 | - | 10 | 900 | |||
AK159212.1 | - | 50 | 3700 | |||
AK159408.1 | - | 50 | 3696 | |||
AK159578.1 | - | 50 | 3699 | |||
AK159958.1 | - | 29 | 2319 | |||
AK160098.1 | - | 50 | 3700 | |||
AK160787.1 | - | 50 | 3695 | |||
AK160788.1 | - | 50 | 3694 | |||
AK160790.1 | - | 50 | 3695 | |||
AK160814.1 | - | 42 | 3164 | |||
AK160946.1 | - | 50 | 3698 | |||
AK161017.1 | - | 50 | 3700 | |||
AK161605.1 | - | 50 | 3695 | |||
AK164628.1 | - | 29 | 2318 | |||
AK164808.1 | - | 50 | 3695 | |||
BC007158.1 | - | 50 | 3679 | 3487477 | ||
BC012438.1 | - | 11 | 1139 | |||
BC042503.1 | - | 50 | 3687 | 3586272 | ||
X58251.1 | - | 50 | 3638 |