Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293559.1 | NP_001280488.1 | 4 | 312 | ||
NM_009941.3 | NP_034071.2 | 4 | 387 | |||
GenBank mRNA | AK002943.1 | - | 644 | |||
AK012583.1 | - | 4 | 324 | |||
AK019276.1 | - | 4 | 289 | |||
AK150447.1 | - | 4 | 317 | |||
AV469576.1 | - | 4 | 312 | |||
BC132269.1 | - | 4 | 308 | 40130546 | ||
BC132275.1 | - | 4 | 308 | 40130552 | ||
M37829.1 | - | 4 | 310 | |||
X54691.1 | - | 4 | 312 |