Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013495.2 | NP_038523.2 | 6 | 747 | ||
GenBank mRNA | AF017175.1 | - | 635 | |||
AK050213.1 | - | 6 | 1040 | |||
AK077454.1 | - | 6 | 701 | |||
AK128925.1 | - | 6 | 701 | |||
AK133472.1 | - | 6 | 702 | |||
AK136487.1 | - | 6 | 701 | |||
AK147770.1 | - | 6 | 702 | |||
AK149688.1 | - | 6 | 698 | |||
BC038395.1 | - | 5 | 682 | 2649352 | ||
BC046383.1 | - | 5 | 682 | 4160676 | ||
BC054791.1 | - | 6 | 728 | 6832731 | ||
D49472.1 | - | 5 | 674 |