Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSN2 SIRNA Assays ›
Gene Symbol:
Csn2
Gene Name:
casein beta
Gene Aliases:
Csnb
Chromosome Location:
Chr.5: 87692619 - 87699429 on Build GRCm38
UniGene ID:
Mm.268737
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s64526
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Csn2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286020.1
NP_001272949.1
6
191
NM_001286021.1
NP_001272950.1
6
206
NM_001286022.1
NP_001272951.1
5
167
NM_001286023.1
NP_001272952.1
4
149
NM_001286024.1
NP_001272953.1
4
146
NM_009972.2
NP_034102.1
6
194
GenBank mRNA
AK021324.1
-
6
188
AK021328.1
-
6
188
AK052803.1
-
6
191
AK052805.1
-
6
191
AK052822.1
-
6
188
AK085678.1
-
6
188
AK085694.1
-
6
191
AK085698.1
-
6
188
AK085729.1
-
6
191
AK085756.1
-
6
188
AK142616.1
-
6
191
AK142620.1
-
6
191
AK142631.1
-
6
191
AK142635.1
-
6
191
AK164747.1
-
6
191
AK164754.1
-
2
515
AK164761.1
-
6
191
AK164791.1
-
6
191
AK164792.1
-
6
188
AK164793.1
-
6
188
AK164794.1
-
6
191
BC003863.1
-
1
16
BC003976.1
-
2
40
BC013332.1
-
6
191
3670912
BC019114.1
-
6
172
3483918
BC019189.1
-
5
184
5028194
BC020356.1
-
6
171
BC020383.1
-
6
171
BC021153.1
-
6
189
3670372
BC050270.1
-
6
171
BC057671.1
-
3
1983
BC080709.1
-
6
175
3671502
BC092098.1
-
6
168
X04490.1
-
6
190
Back To Top
More Information
Panther Classification:
Molecular Function -
storage protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
calcium ion transport
lactation
negative regulation of catalytic activity
negative regulation of lactation
enzyme inhibitor activity
transporter activity
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0