Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007790.3 | NP_031816.2 | 8 | 649 | ||
GenBank mRNA | AF047601.1 | - | 8 | 576 | ||
AF141294.1 | - | 8 | 599 | |||
AK140705.1 | - | 8 | 634 | |||
AK143996.1 | - | 8 | 638 | |||
AK164387.1 | - | 8 | 642 | |||
BC062935.1 | - | 8 | 625 | 6838083 |