Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007799.3 | NP_031825.2 | 3 | 505 | ||
GenBank mRNA | AK143581.1 | - | 2 | 207 | ||
AK145875.1 | - | 3 | 362 | |||
AK157907.1 | - | 2 | 227 | |||
AK165242.1 | - | 2 | 227 | |||
AK165271.1 | - | 2 | 173 | |||
BC005432.1 | - | 3 | 505 | 3601519 | ||
CT010252.1 | - | 2 | 141 | |||
X97399.1 | - | 2 | 192 |