Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 11 | 1434 | ||
NM_001291234.1 | NP_001278163.1 | 11 | 1139 | |||
NM_001291238.1 | NP_001278167.1 | 11 | 1145 | |||
NM_001291239.1 | NP_001278168.1 | 11 | 1428 | |||
NM_001291240.1 | NP_001278169.1 | 11 | 1434 | |||
NM_009986.4 | NP_034116.3 | 11 | 1145 | |||
NM_198602.3 | NP_941004.2 | 11 | 1139 | |||
GenBank mRNA | AB041595.1 | - | 11 | 1278 | ||
AF004225.1 | - | 7 | 498 | |||
AK098096.1 | - | 11 | 902 | |||
AK144932.1 | - | 11 | 1799 | |||
AK220173.1 | - | 11 | 895 | |||
AY037807.1 | - | 11 | 875 | |||
BC014289.1 | - | 11 | 914 | |||
BC054370.1 | - | 11 | 862 | |||
BC079570.1 | - | 12 | 1332 | |||
BC090847.1 | - | 11 | 917 | 30547458 | ||
U68542.1 | - | 12 | 913 | |||
X75013.1 | - | 7 | 629 |