Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 2, 3 | 721 | ||
NM_001291234.1 | NP_001278163.1 | 2, 3 | 432 | |||
NM_001291238.1 | NP_001278167.1 | 2, 3 | 432 | |||
NM_001291239.1 | NP_001278168.1 | 2, 3 | 721 | |||
NM_001291240.1 | NP_001278169.1 | 2, 3 | 721 | |||
NM_009986.4 | NP_034116.3 | 2, 3 | 432 | |||
NM_198602.3 | NP_941004.2 | 2, 3 | 432 | |||
GenBank mRNA | AB041595.1 | - | 2, 3 | 568 | ||
AK098096.1 | - | 2, 3 | 195 | |||
AK133543.1 | - | 2, 3 | 195 | |||
AK144932.1 | - | 2, 3 | 1092 | |||
AK220173.1 | - | 2, 3 | 188 | |||
AY037807.1 | - | 2, 3 | 162 | |||
BC014289.1 | - | 2, 3 | 201 | |||
BC054370.1 | - | 2, 3 | 155 | |||
BC079570.1 | - | 3, 4 | 619 | |||
BC090847.1 | - | 2, 3 | 204 | 30547458 | ||
CA317682.1 | - | 2, 3 | 387 | |||
CJ177481.1 | - | 2, 3 | 349 | |||
U68542.1 | - | 3, 4 | 206 |