Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109750.1 | NP_001103220.1 | 7 | 1131 | ||
NM_010048.3 | NP_034178.2 | 7 | 1140 | |||
GenBank mRNA | AK042786.1 | - | 7 | 1062 | ||
AK129070.1 | - | 8 | 990 | |||
AK133880.1 | - | 4 | 429 | |||
AK141594.1 | - | 7 | 1106 | |||
AK145495.1 | - | 7 | 1057 | |||
AK153026.1 | - | 7 | 928 | |||
AK161022.1 | - | 1891 | ||||
AK161359.1 | - | 7 | 856 | |||
AK163872.1 | - | 7 | 1093 | |||
AK163903.1 | - | 7 | 1069 | |||
AK190458.1 | - | 2 | 191 | |||
AK210025.1 | - | 1 | 72 | |||
BC060636.1 | - | 7 | 1081 | 6849298 | ||
BC062978.1 | - | 7 | 1035 | 30356000 | ||
D78641.1 | - | 7 | 1065 | |||
X95480.1 | - | 7 | 1006 |