Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109750.1 | NP_001103220.1 | 10 | 1773 | ||
NM_010048.3 | NP_034178.2 | 10 | 1782 | |||
GenBank mRNA | AK088131.1 | - | 9 | 1499 | ||
AK129070.1 | - | 11 | 1632 | |||
AK133880.1 | - | 7 | 1071 | |||
AK141594.1 | - | 10 | 1748 | |||
AK145495.1 | - | 10 | 1699 | |||
AK153026.1 | - | 10 | 1569 | |||
AK161022.1 | - | 2533 | ||||
AK161359.1 | - | 10 | 1498 | |||
AK163872.1 | - | 10 | 1735 | |||
AK163903.1 | - | 10 | 1711 | |||
AK164263.1 | - | 3 | 482 | |||
BC031506.1 | - | 3 | 492 | |||
BC060636.1 | - | 10 | 1723 | 6849298 | ||
BC062978.1 | - | 10 | 1677 | 30356000 | ||
D78641.1 | - | 10 | 1707 | |||
X95480.1 | - | 10 | 1648 |