Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306146.1 | NP_001293075.1 | 11 | 1498 | ||
NM_002112.3 | NP_002103.2 | 12 | 1597 | |||
XM_017022094.1 | XP_016877583.1 | 12 | 1466 | |||
XM_017022095.1 | XP_016877584.1 | 11 | 1367 | |||
XM_017022096.1 | XP_016877585.1 | 11 | 1251 | |||
XM_017022097.1 | XP_016877586.1 | 10 | 1549 | |||
XM_017022098.1 | XP_016877587.1 | 9 | 1069 | |||
GenBank mRNA | AK027221.1 | - | 5 | 1000 | ||
BC130527.1 | - | 12 | 1298 | 40146558 | ||
BC144173.1 | - | 11 | 1199 | 9052693 | ||
M60445.1 | - | 12 | 1306 | |||
X54297.1 | - | 12 | 1357 |