Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010056.3 | NP_034186.2 | 1 | 319 | ||
NM_198854.2 | NP_942151.1 | 1 | 319 | |||
GenBank mRNA | AF022075.1 | - | 1 | 161 | ||
AF022076.1 | - | 1 | 161 | |||
AF022077.1 | - | 1 | 161 | |||
AF033011.1 | - | 1 | 159 | |||
AF072452.1 | - | 1 | 319 | |||
AF072453.1 | - | 1 | 319 | |||
AK134920.1 | - | 1 | 195 | |||
AK158121.1 | - | 1 | 334 | |||
AK158772.1 | - | 1 | 254 | |||
U67840.1 | - | 2 | 295 |