Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007870.3 | NP_031896.3 | 3 | 489 | ||
GenBank mRNA | AF047355.1 | - | 3 | 464 | ||
AK156109.1 | - | 322 | ||||
AK171843.1 | - | 3 | 322 | |||
AK172254.1 | - | 3 | 882 | |||
BC012671.1 | - | 3 | 442 | 4160709 | ||
U76110.1 | - | 3 | 298 |